2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxy-5-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(2-methoxy-5-nitro-phenoxy)-5-(trifluoromethyl)benzene

PC502760-1G
APOSPC502760-1GEA 1180 EUR
APOSPC502760-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxy-5-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxy-5-nitrophenyl ether
Formel: C₁₄H₈F₅NO₄
Molecular Weight: 349,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630053
CAS-Nummer: 2244087-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR