2,6-Difluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2,6-Difluoro-4-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502761-1G
APOSPC502761-1GEA 1180 EUR
APOSPC502761-1G
2,6-Difluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,6-Difluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄F₇NO₃
Molecular Weight: 355,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630056
CAS-Nummer: 2244083-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR