2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methyl-5-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-fluoro-2-methyl-5-nitrobenzene

PC502762-1G
APOSPC502762-1GEA 1180 EUR
APOSPC502762-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methyl-5-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methyl-5-nitrophenyl ether
Formel: C₁₄H₇F₆NO₃
Molecular Weight: 351,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630059
CAS-Nummer: 2244083-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR