4,5-Dichloro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(4,5-Dichloro-2-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502764-1G
APOSPC502764-1GEA
1180
EUR
APOSPC502764-1G
4,5-Dichloro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4,5-Dichloro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₄Cl₂F₅NO₃ Molecular Weight: 388,074 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630065 CAS-Nummer: 2244083-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...