4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromo-2,6-dimethyl-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502767-1G
APOSPC502767-1GEA 1180 EUR
APOSPC502767-1G
4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₅H₁₀BrF₅O
Molecular Weight: 381,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630074
CAS-Nummer: 2244084-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR