4-Bromo-3,5-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-1,3-dimethylbenzene

PC502768-1G
APOSPC502768-1GEA 1180 EUR
APOSPC502768-1G
4-Bromo-3,5-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-3,5-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₅H₁₀BrF₅O
Molecular Weight: 381,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630077
CAS-Nummer: 2244087-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR