2-Bromo-4-(2,4,4-trimethylpentan-2-yl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2-Bromo-4-(1,1,3,3-tetramethylbutyl)phenoxy]-1,3-difluoro-5-(trifluoromethyl)benzene

PC502770-1G
APOSPC502770-1GEA 1180 EUR
APOSPC502770-1G
2-Bromo-4-(2,4,4-trimethylpentan-2-yl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-(2,4,4-trimethylpentan-2-yl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₂₁H₂₂BrF₅O
Molecular Weight: 465,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630083
CAS-Nummer: 2244087-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR