5-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Bromo-2,3-difluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502771-1G
APOSPC502771-1GEA 1180 EUR
APOSPC502771-1G
5-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₇O
Molecular Weight: 389,063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630086
CAS-Nummer: 2244086-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR