6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-3-fluoro-4-(trifluoromethoxy)benzene

PC502772-1G
APOSPC502772-1GEA 1180 EUR
APOSPC502772-1G
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₄BrF₉O₂
Molecular Weight: 455,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630089
CAS-Nummer: 2244085-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR