2-Bromo-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-4-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502774-1G
APOSPC502774-1GEA 1180 EUR
APOSPC502774-1G
2-Bromo-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈BrF₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630095
CAS-Nummer: 2244085-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR