2-Bromo-4-(tert-butyl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502775-1G
APOSPC502775-1GEA 1180 EUR
APOSPC502775-1G
2-Bromo-4-(tert-butyl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-(tert-butyl)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₇H₁₄BrF₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD31630098
CAS-Nummer: 2244088-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR