5-Bromo-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502776-1G
APOSPC502776-1GEA 1180 EUR
APOSPC502776-1G
5-Bromo-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2,4-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₇O
Lagertemperatur: Raumtemperatur
MDL: MFCD31630101
CAS-Nummer: 2244085-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR