5-Bromo-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-4-fluoro-2-methylbenzene

PC502778-1G
APOSPC502778-1GEA 1180 EUR
APOSPC502778-1G
5-Bromo-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2-fluoro-4-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₇BrF₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD31629956
CAS-Nummer: 2244084-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR