2-Bromo-6-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-6-methyl-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502779-1G
APOSPC502779-1GEA 1180 EUR
APOSPC502779-1G
2-Bromo-6-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-6-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈BrF₅O
Molecular Weight: 367,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629960
CAS-Nummer: 2244088-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR