4-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Bromo-2-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502780-1G
APOSPC502780-1GEA
1180
EUR
APOSPC502780-1G
4-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₅BrF₆O Molecular Weight: 371,073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629964 CAS-Nummer: 2244087-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...