5-Bromo-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Bromo-2-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502781-1G
APOSPC502781-1GEA 1180 EUR
APOSPC502781-1G
5-Bromo-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈BrF₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31629967
CAS-Nummer: 2244083-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR