4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromo-3-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502782-1G
APOSPC502782-1GEA 1180 EUR
APOSPC502782-1G
4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈BrF₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31629970
CAS-Nummer: 2244086-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR