2-Bromo-6-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(2-Bromo-6-iodo-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502783-1G
APOSPC502783-1GEA
1180
EUR
APOSPC502783-1G
2-Bromo-6-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-6-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₅BrF₅IO Molecular Weight: 478.979 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629973 CAS-Nummer: 2244086-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...