2-Bromo-4,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-3,5-dimethylbenzene

PC502784-1G
APOSPC502784-1GEA 1180 EUR
APOSPC502784-1G
2-Bromo-4,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₅H₁₀BrF₅O
Molecular Weight: 381,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629976
CAS-Nummer: 2244086-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR