5-Bromo-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Bromo-2-methyl-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502785-1G
APOSPC502785-1GEA 1180 EUR
APOSPC502785-1G
5-Bromo-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈BrF₅O
Molecular Weight: 367,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629979
CAS-Nummer: 2244086-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR