2-Bromo-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-5-iodo-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502786-1G
APOSPC502786-1GEA 1180 EUR
APOSPC502786-1G
2-Bromo-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅IO
Molecular Weight: 478,979 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629982
CAS-Nummer: 2244084-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR