3-Bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
PC502787-1G
APOSPC502787-1GEA
1180
EUR
APOSPC502787-1G
3-Bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Bromo-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₅BrF₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629985 CAS-Nummer: 2244084-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...