3-Bromo-2-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(3-Bromo-2-iodo-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502788-1G
APOSPC502788-1GEA
1180
EUR
APOSPC502788-1G
3-Bromo-2-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Bromo-2-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₅BrF₅IO Molecular Weight: 478,979 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629988 CAS-Nummer: 2244088-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...