2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2-Bromo-4-(trifluoromethoxy)phenoxy]-1,3-difluoro-5-(trifluoromethyl)benzene

PC502789-1G
APOSPC502789-1GEA 1180 EUR
APOSPC502789-1G
2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₅BrF₈O₂
Molecular Weight: 437,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629991
CAS-Nummer: 2244087-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR