5-Bromo-2-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Bromo-2-chloro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502791-1G
APOSPC502791-1GEA 1180 EUR
APOSPC502791-1G
5-Bromo-2-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-2-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrClF₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD31629997
CAS-Nummer: 2244086-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR