4-Bromo-3,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-1,3-difluorobenzene

PC502792-1G
APOSPC502792-1GEA 1180 EUR
APOSPC502792-1G
4-Bromo-3,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-3,5-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₇O
Molecular Weight: 389,063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630000
CAS-Nummer: 2244084-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR