5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-5-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-2-fluoro-4-methylbenzene

PC502794-1G
APOSPC502794-1GEA 1180 EUR
APOSPC502794-1G
5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₇BrF₆O
Molecular Weight: 385,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630006
CAS-Nummer: 2244086-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR