3-Bromo-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Bromo-4-iodo-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502796-1G
APOSPC502796-1GEA 1180 EUR
APOSPC502796-1G
3-Bromo-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Bromo-4-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅IO
Molecular Weight: 478.979 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630012
CAS-Nummer: 2244087-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR