2-Bromo-6-nitro-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-[2-Bromo-6-nitro-4-(trifluoromethoxy)phenoxy]-1,3-difluoro-5-(trifluoromethyl)benzene

PC502798-1G
APOSPC502798-1GEA 1180 EUR
APOSPC502798-1G
2-Bromo-6-nitro-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-6-nitro-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₄BrF₈NO₄
Molecular Weight: 482,077 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630018
CAS-Nummer: 2244086-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR