2-Chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Chloro-4-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502799-1G
APOSPC502799-1GEA 1180 EUR
APOSPC502799-1G
2-Chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅ClF₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD31630021
CAS-Nummer: 2244084-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR