2,6-Difluoro-4-(trifluoromethyl)phenyl 4-ethyl-5-fluoro-2-methoxyphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-ethyl-5-fluoro-2-methoxybenzene

PC502800-1G
APOSPC502800-1GEA 1180 EUR
APOSPC502800-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-ethyl-5-fluoro-2-methoxyphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-ethyl-5-fluoro-2-methoxyphenyl ether
Formel: C₁₆H₁₂F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630024
CAS-Nummer: 2244087-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR