2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluorophenyl ether 95%
Lieferant: Apollo Scientific
PC502801-1G
APOSPC502801-1GEA
1180
EUR
APOSPC502801-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluorophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluorophenyl ether
Formel:
C₁₃H₆F₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630027 CAS-Nummer: 2244087-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...