2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-methoxyphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(5-fluoro-2-methoxy-phenoxy)-5-(trifluoromethyl)benzene

PC502802-1G
APOSPC502802-1GEA 1180 EUR
APOSPC502802-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-methoxyphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-methoxyphenyl ether
Formel: C₁₄H₈F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630030
CAS-Nummer: 2244087-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR