2-Chloro-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(2-Chloro-6-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502804-1G
APOSPC502804-1GEA
1180
EUR
APOSPC502804-1G
2-Chloro-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₅ClF₆O Molecular Weight: 326,622 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630036 CAS-Nummer: 2244084-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...