2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methoxyphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(4-fluoro-2-methoxy-phenoxy)-5-(trifluoromethyl)benzene

PC502805-1G
APOSPC502805-1GEA 1180 EUR
APOSPC502805-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methoxyphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-2-methoxyphenyl ether
Formel: C₁₄H₈F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630039
CAS-Nummer: 2244087-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR