4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Chloro-3-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502806-1G
APOSPC502806-1GEA 1180 EUR
APOSPC502806-1G
4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅ClF₆O
Molecular Weight: 326,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630042
CAS-Nummer: 2244086-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR