2,4-Dibromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
PC502807-1G
APOSPC502807-1GEA
1180
EUR
APOSPC502807-1G
2,4-Dibromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,4-Dibromo-6-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₄Br₂F₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630045 CAS-Nummer: 2244087-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...