2,6-Dichloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Dichloro-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-5-fluorobenzene
PC502809-1G
APOSPC502809-1GEA
1180
EUR
APOSPC502809-1G
2,6-Dichloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,6-Dichloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₄Cl₂F₆O Molecular Weight: 361,067 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630051 CAS-Nummer: 2244084-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...