2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluorophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-2-(2-fluorophenoxy)-5-(trifluoromethyl)benzene
PC502811-1G
APOSPC502811-1GEA
1180
EUR
APOSPC502811-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluorophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluorophenyl ether
Formel:
C₁₃H₆F₆O Molecular Weight: 292,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630057 CAS-Nummer: 2244086-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...