2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(2-fluoro-4-methyl-phenoxy)-5-(trifluoromethyl)benzene

PC502812-1G
APOSPC502812-1GEA 1180 EUR
APOSPC502812-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether
Formel: C₁₄H₈F₆O
Molecular Weight: 306,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630060
CAS-Nummer: 2244084-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR