3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Chloro-2-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502814-1G
APOSPC502814-1GEA 1180 EUR
APOSPC502814-1G
3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅ClF₆O
Molecular Weight: 326,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630066
CAS-Nummer: 2244086-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR