2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-2-iodophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(3-fluoro-2-iodo-phenoxy)-5-(trifluoromethyl)benzene

PC502815-1G
APOSPC502815-1GEA 1180 EUR
APOSPC502815-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-2-iodophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-2-iodophenyl ether
Formel: C₁₃H₅F₆IO
Molecular Weight: 418,073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630069
CAS-Nummer: 2244087-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR