2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-2-(5-fluoro-2-iodo-phenoxy)-5-(trifluoromethyl)benzene
PC502816-1G
APOSPC502816-1GEA
1180
EUR
APOSPC502816-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether
Formel:
C₁₃H₅F₆IO Molecular Weight: 418,073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630072 CAS-Nummer: 2244087-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...