2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502817-1G
APOSPC502817-1GEA 1180 EUR
APOSPC502817-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether
Formel: C₁₄H₅F₉O
Lagertemperatur: Raumtemperatur
MDL: MFCD31630075
CAS-Nummer: 2244086-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR