2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-iodo-2-methylphenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(4-fluoro-5-iodo-2-methyl-phenoxy)-5-(trifluoromethyl)benzene

PC502818-1G
APOSPC502818-1GEA 1180 EUR
APOSPC502818-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-iodo-2-methylphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-iodo-2-methylphenyl ether
Formel: C₁₄H₇F₆IO
Molecular Weight: 432,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630078
CAS-Nummer: 2244086-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR