2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1-Chloro-2-[2,6-difluoro-4-(trifluoromethyl)phenoxy]-5-fluoro-4-iodobenzene
PC502819-1G
APOSPC502819-1GEA
1180
EUR
APOSPC502819-1G
2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₄ClF₆IO Molecular Weight: 452,518 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630081 CAS-Nummer: 2244084-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...