2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-methylphenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-2-(3-fluoro-4-methyl-phenoxy)-5-(trifluoromethyl)benzene
PC502820-1G
APOSPC502820-1GEA
1180
EUR
APOSPC502820-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-methylphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-methylphenyl ether
Formel:
C₁₄H₈F₆O Molecular Weight: 306,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630084 CAS-Nummer: 2244085-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...