2-Chloro-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Chloro-5-fluoro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502821-1G
APOSPC502821-1GEA 1180 EUR
APOSPC502821-1G
2-Chloro-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅ClF₆O
Molecular Weight: 326,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630087
CAS-Nummer: 2244087-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR