2-Chloro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Chloro-4-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502823-1G
APOSPC502823-1GEA 1180 EUR
APOSPC502823-1G
2-Chloro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-4-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈ClF₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31630093
CAS-Nummer: 2244084-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR