5-Chloro-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Chloro-2-methoxy-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502824-1G
APOSPC502824-1GEA 1180 EUR
APOSPC502824-1G
5-Chloro-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Chloro-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₄H₈ClF₅O₂
Molecular Weight: 338,657 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630096
CAS-Nummer: 2244086-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR